Learn More
Medchemexpress LLC HY-17400 100mg Medchemexpress, Tegafur CAS:17902-23-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY17400100MG
Encompass_Preferred
Medchemexpress, HY-17400 100mg Tegafur CAS:17902-23-7 Tegafur (FT 207; NSC 148958) is a chemotherapeutic 5-FU prodrug used in the treatment of cancers; is a component of tegafur-uracil. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1944