Learn More
Selleck Chemical LLC Tegaserod E4976-100MG
SDP
Supplier:  Selleck Chemical LLC E4976100MG
Encompass_Preferred
Tegaserod is an orally active agonist of serotonin receptor 4 (5-HT4R) and also acts as an antagonist of 5-HT2B receptor with pKi values of 7 5 8 4 and 7 0 for human recombinant 5-HT2A 5-HT2B and 5-HT2C receptors respectively Tegaserod has anti-tumor activity and is used in the research of irritable bowel syndrome (IBS)
Catalog No. 50-000-41629