Learn More
Cayman Chemical Tegaserodmaleate 10mg
SDP
Supplier:  Cayman Chemical 1469210 MG
Encompass_Preferred
A potent agonist of the serotonin 4 receptor (5-HT4, Ki = 3.9-31 nM; also acts as an antagonist for the 5-HT2A, 5-HT2B, and 5-HT2C receptors (Ki = 32, 3.9, and 100 nM, respectively); stimulates colonic motility and transit in vivo
Catalog No. 50-297-2993