Learn More
Medchemexpress LLC HY-109103 5mg Medchemexpress, Tegatrabetan CAS:1227637-23-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1091035MG
Encompass_Preferred
Medchemexpress, HY-109103 5mg Tegatrabetan CAS:1227637-23-1 Tegatrabetan (BC2059) is a β-Catenin antagonist. Tegatrabetan disrupts the binding of β-catenin with the scaffold protein transducin β-like 1 (TBL1). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9664