Learn More
TARGETMOL CHEMICALS INC Tegobuvir 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T170295MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Tegobuvir (GS 333126) is a specific and covalent HCV NS5B polymerase inhibitor. Purity 99.86%
Catalog No. 50-287-7401