Learn More
Medchemexpress LLC HY-10235 10mg Medchemexpress, Telaprevir CAS:402957-28-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1023510MG
Encompass_Preferred
Medchemexpress, HY-10235 10mg Telaprevir CAS:402957-28-2 Telaprevir is a highly selective, reversible, and potent peptidomimetic inhibitor of the HCV NS3-4A protease, the steady-state inhibitory constant (Ki) of Telaprevir is 7 nM against a genotype 1 (H strain) NS3 protease domain plus a NS4A cofactor peptide. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7984