Learn More
Apexbio Technology LLC Telaprevir (VX-950), 25mg. Cas: 402957-28-2 MFCD: MFCD11616089. HCV NS3-4A protease inhibitor
SDP
Supplier:  Apexbio Technology LLC A403125
Encompass_Preferred
Telaprevir (also known as VX-950), derived from the viral NS5A/5B substrate of the protease through structure-based techniques, is a novel and potent inhibitor of hepatitis C virus (HCV) NS3-4A protease 402957-28-2. MFCD11616089
Catalog No. 50-101-3647