Learn More
TARGETMOL CHEMICALS INC Telatinib 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T616650MG
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg and bulk. Please contact Fisher for quotes. Telatinib (Bay 57-9352) is an effective inhibitor of VEGFR2/3, c-Kit, and PDGFRalpha, with IC50s of 6 nM/4 nM, 1 nM, and 15 nM, respectively. Purity 98.95%
Catalog No. 50-288-0022