Learn More
Medchemexpress LLC Telisotuzumab (Synonyms: ABT-700) 10mg
SDP
Supplier:  Medchemexpress LLC HYP9939110MG
Encompass_Preferred
Telisotuzumab (ABT-700) is a human recombinant bivalent antibody, a therapeutic antibody against the hepatocyte growth factor receptor (MET) that binds c-Met with high affinity and inhibits c-Met signaling. Telisotuzumab has antitumor activity.
Catalog No. NC3397890