Learn More
Medchemexpress LLC Tellimagrandin II (Eugeniin) | 81571-72-4 | 98.8% | 938.66 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN93865MG
Encompass_Preferred
Tellimagrandin II (Eugeniin) is the first intermediate of the ellagitannin series, derived from 4C1-glucose, and possesses oral activity. It has shown potential for research in antibacterial, anti-inflammatory, and neurodegenerative diseases due to its diverse biological effects.
  • Inhibits resistance of Staphylococcus aureus by disrupting cell wall integrity.
  • Exhibits anti-inflammatory effects by inhibiting NO production and COX-2 levels in macrophages.
  • Improves memory impairment by inhibiting acetylcholinesterase (AChE) activity.
  • Demonstrates improved learning and memory functions in scopolamine-induced amnesiac mice.
Catalog No. 50-004-25349