Learn More
TARGETMOL CHEMICALS INC Telmisartan 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1570200MG
Encompass_Preferred
Also available in 1 g, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Telmisartan (BIBR 277) is an Angiotensin 2 Receptor Blocker. The mechanism of action of telmisartan is as an Angiotensin 2 Receptor Antagonist. Purity 98.78%
Catalog No. 50-287-8473