Learn More
Medchemexpress LLC Telmisartan-d3 | 1189889-44-8 | >99.7% | 517.64 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY13955S1MG
Encompass_Preferred
Telmisartan-d3 is a deuterium-labeled version of Telmisartan, a potent and long-lasting antagonist of the angiotensin II type 1 receptor (AT1). It selectively inhibits the binding of 125I-AngII to AT1 receptors with an IC50 of 9.2 nM.
  • Intended for research use only
  • Can be utilized as a tracer or internal standard
  • Suitable for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS
  • Deuteration influences pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-49542