Learn More
Medchemexpress LLC Telotristat ethyl | 1033805-22-9 | 98.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY13055A100MG
Encompass_Preferred
Telotristat ethyl is a novel, orally-delivered inhibitor of tryptophan hydroxylase that reduces serotonin production. It is intended for research use only.
  • Reduces serotonin content in the periphery.
  • Prevents increase in blood neutrophil counts after TNBS challenge.
  • Provides significant protection in a mouse model of inflammatory bowel disease.
  • Protects mouse IBD model, confirmed by histopathology evaluation.
  • Depletes 5-HT from the jejunum.
  • Alleviates the severity of trinitrobenzene sulfonic acid (TNBS)-induced colitis.
Catalog No. 50-003-38371