Learn More
Simpore Inc 500×500 µm opening2.9 mm diameter + 0.1 mm thick chip framePack of 10
Supplier:  Simpore Inc APT100Q01
Encompass_Preferred
500×500 µm opening2.9 mm diameter + 0.1 mm thick chip framePack of 10
Catalog No. NC2818824