Learn More
Research Products International Corp TEMED [N,N,N',N'-Tetramethylethylenediamine], 100 Milliliters
SDP
Supplier:  Research Products International Corp T180000.1
Encompass_Preferred
Tertiary amine used to catalyze formation of free radicals from ammonium persulfate and riboflavin. Suitable for electrophoresis and tested as a catalyst for polyacrylamide gel electrophoresis.
Catalog No. 50-489-060