Learn More
Medchemexpress LLC Temocapril | 111902-57-9 | MFCD00865951 | 98.1% | 476.61 g/mol | C23H28N2O5S2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1007135MG
Encompass_Preferred
Temocapril is an orally active angiotensin-converting enzyme (ACE) inhibitor offered as a research reagent for studies of hypertension, congestive heart failure, and related cardiovascular conditions. Supplied as a solid powder for laboratory use only; not for human or veterinary use.
  • Orally active ACE inhibitor.
  • Used in hypertension and cardiovascular research.
  • Available in small sample sizes for analytical and preparative work.
  • High purity (≈98.1%).
  • Molecular weight 476.61 g/mol, formula C23H28N2O5S2.
  • Supplied as a powder suitable for storage and analytical testing.
Catalog No. 50-002-14517