Learn More
Apexbio Technology LLC Temocapril HCl 110221-44-8 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B22135.1
Encompass_Preferred
Temocapril HCl (CAS 110221-44-8) is an orally administered angiotensin-converting enzyme (ACE) inhibitor that suppresses the enzymatic conversion of angiotensin I to the vasoconstrictor angiotensin II thereby reducing vascular tone and aldosterone secretion Through modulation of the renin-angiotensin-aldosterone system Temocapril HCl is utilized in biomedical research to investigate mechanisms underlying hypertension congestive heart failure acute myocardial infarction insulin resistance and chronic kidney disease as well as to evaluate pharmacological interventions targeting these conditions
Catalog No. 50-000-90649