Learn More
TARGETMOL CHEMICALS INC Temozolomide 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1178200MG
Encompass_Preferred
Also available in 1 g, 1 mL, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Temozolomide (TMZ) is a DNA alkylating agent with blood-brain barrier permeability and oral activity. Temozolomide has antitumor activity and antiangiogenic activity, and also induces apoptosis and autophagy. Temozolomide is stable under acidic conditions and hydrolyzes under neutral or slightly alkaline conditions. Purity 99.87%
Catalog No. 50-287-5909