Learn More
Medchemexpress LLC HY-17364 1g , Temozolomide NSC 362856;CCRG 81045;TZM CAS:85622-93-1 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY173641G
Encompass_Preferred
Medchemexpress, HY-17364 1g Temozolomide NSC 362856;CCRG 81045;TZM CAS:85622-93-1 Formula:C6H6N6O2 DNA alkylator Purity:98% Temozolomide is a DNA-methylating agent. Temozolomide is an alkylating cytostatic drug. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2283