Learn More
Medchemexpress LLC Temozolomide acid | 113942-30-6 | MFCD12032160 | 99.7% | 195.14 g/mol | C6H5N5O3 | 100 MG

Supplier: Medchemexpress LLC HYW018326100MG
Temozolomide acid is a carboxylic acid derivative of temozolomide used as a research anticancer agent. It functions as a DNA alkylating agent that can induce DNA strand breaks, cell cycle arrest, and cell death, and is commonly used in glioblastoma and brain cancer research.
- Carboxylic acid derivative of temozolomide.
- Acts as a DNA alkylating agent causing DNA breaks and cell cycle arrest.
- Relevant for glioblastoma and brain cancer research.
- Molecular weight 195.14 g/mol.
- Chemical formula C6H5N5O3.
- High purity per manufacturer specifications.
- Supplied as a solid research material, commonly in 100 mg packs.
- Storage: powder -20°C (long term) or 4°C (short term).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.