Learn More
Medchemexpress LLC Temozolomide acid | 113942-30-6 | MFCD12032160 | 99.7% | 195.14 g/mol | C6H5N5O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW01832650MG
Encompass_Preferred
Temozolomide acid is the carboxylic acid derivative of temozolomide (CAS 113942-30-6) used as a research reagent in anticancer studies. It is applied to investigations of DNA/RNA synthesis and alkylation mechanisms and is supplied as a high-purity solid for in vitro and in vivo experimental work.
  • Carboxylic acid derivative of temozolomide.
  • Used in anticancer and DNA/RNA synthesis research.
  • High purity suitable for analytical and biological assays.
  • Supplied in small research-scale packages for laboratory use.
  • Molecular formula C6H5N5O3; molecular weight 195.14 g/mol.
Catalog No. 50-003-01155