Learn More
Cayman Chemical TemozolomIde AcId 25mg
SDP
Supplier:  Cayman Chemical 3580025 MG
Encompass_Preferred
An active metabolite of temozolomide; cytotoxic to TLX5 murine lymphoma cells; a synthetic intermediate in the synthesis of temozolomide prodrugs with anticancer activity
Catalog No. 50-291-1406