Learn More
Selleck Chemical LLC Temozolomide 25mg 85622-93-1 CCRG81045, NSC 362856,TMZ
SDP
Supplier:  Selleck Chemical LLC S123725MG
Encompass_Preferred
Temozolomide (CCRG81045, NSC 362856, TMZ) is a monofunctional SN-1 alkylating agent that can modify nitrogen atoms in the DNA ring and the extracyclic oxygen group, chemically converted to MTIC and degrades to methyldiazonium cation, which transfers methyl groups to DNA at physiologic pH. A DNA damage inducer in L-1210 and L-1210/BCNU cells. Temozolomide induces apoptosis and exhibits antitumor activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-711-6