Learn More
Cayman Chemical TemozolomIde-d3 1mg
SDP
Supplier:  Cayman Chemical 223721 MG
Encompass_Preferred
An internal standard for the quantification of temozolomide by GC- or LC-MS
Catalog No. 50-298-7764