Learn More
Saf T Pak Inc Sepio Ambient Insulated Shipping System, 75-28L, 15° to 25°C
SDP
Supplier:  Saf T Pak Inc 20627
Encompass_Preferred
The cost-effective SEPIO is a tested system offering a low barrier of entry when shipping payloads needing refrigeration, and can be used as an Overpack when transporting Category A Infectious Substances (UN 2814 or UN 2900).FEATURES: Key Features | ClaimsPre-qualified for 2°C to 8°C and 15°C to 25°C using the same brick cassettesInmark Legacy ProfilesSeasonal and; Universal pack-outsCorrugated Cassettes for Easy HandlingMultiple payload sizesCost-effective solutionsWater-based refrigerant cassettesGlobal availability
Catalog No. 50-190-4600