Learn More
Saf T Pak Inc Fortis 45-13.3L CRT (15°-25°C) Insulated Shipping Solution (Single Pack)
SDP
Supplier:  Saf T Pak Inc 38475
Encompass_Preferred
FORTIS (2°-8°C, PCM6 and; Neopor)Designed for use when shipping high-risk, irreplaceable payloads, the FORTIS product line is our flagship 2°C to 8°C shipping system. Using a graphite-infused polystyrene-insulated chest, Neopor, and a unique Phase Change Material that phases between 2°C to 8°C, this robust system has been pre-qualified to ISTA 7D.This system can be used as an Overpack when transporting Category A Infectious Substances (UN 2814 or UN 2900). The Fortis shipping solution is a high performance package designed for single use shipments in international lanes. By validating the shippers to the most challenging industry standard profiles, the Fortis is a cost effective solution that takes risk out of your supply chain.FEATURES:Tested to ISTA 7D Profiles2°-8°C Universal Solution: 48 and 72 hour15°-25°C Solution: 48, 72, 96 hourAdvanced Phase Change MaterialPriced For Single Use - Robust Enough for Multiple Shipments
Catalog No. 50-190-4566