Learn More
Analytical Sales & Services TEMPERATURE CONTROLLED REACTOR
Supplier:  Analytical Sales & Services 249800
Encompass
24 Position,18mm Spacing, Temperature Controlled Reactor with Temperature Transfer Cover; Includes Rubber Gasket, Rubber and Viton O-Rings, CPC Fittings and PUR Tubing
Catalog No. NC2891425