Learn More
Research Products International Corp Iso-Chill Flipper Temperature Rack, 0C, White, 7 x 4 1/4 x 4 Inches
SDP
Supplier:  Research Products International Corp 248000
Encompass
Iso-Chill Flipper rack, white 0°C unit will maintain a temperature of 0°C for a minimum of five hours, for keeping enzymes, cells and other temperature sensitive reagents cool and stable when on a benchtop. Also protect samples in event of a freezer or power failure. Iso-Chill Flipper racks can accommodate up to twenty 1.5 ml - 2.0 ml micro-tubes or screw cap tubes on one side or twenty 0.5 ml tubes on the reverse side. Both rack and lid are constructed of virtually unbreakable polycarbonate.
Catalog No. 50-476-533