Learn More
Sigma Aldrich Fine Chemicals Biosciences TempliPhiTM 100 Amplificat
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE25640010
Encompass
illustra(TM) TempliPhi(TM) DNA Amplification Kits use isothermal rolling circle amplification (RCA) to efficiently prepare DNA sequencing templates.TempliPhi(TM) Kits use a unique process to efficiently prepare micrograms of circular DNA from picogram input material. The DNA templates are prepared by rolling circle amplification (RCA) using bacteriophage Phi29 DNA polymerase. TempliPhi(TM) uses an isothermal method for the exponential amplification of circular DNA. Phi29 DNA polymerase is active at 30 C enabling amplification to be performed at this temperature without the need for thermal cycling. The TempliPhi(TM) protocol requires less than 20 min of hands-on time to amplify 96 samples from bacterial colonies.Two different types of TempliPhi(TM) DNA Amplification Kits are available to meet different throughput requirements. illustra(TM) TempliPhi(TM) DNA Amplification Kit for 2000 reactions contains premixed components and generates templates after overnight incubation. The 100- an
Catalog No. 50-290-4485