Learn More
Medchemexpress LLC Tempo | 2564-83-2 | MFCD00009599 | 156.25 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW00118750G
Encompass_Preferred
Tempo is a nitric oxide radical and a selective scavenger of reactive oxygen species (ROS) in mitochondria. It functions as an organocatalyst that disproportionates superoxide and oxidizes primary alcohols to aldehydes in a catalytic cycle. It exhibits mutagenic and antioxidant effects, and can induce DNA strand breaks, as well as exerting cytotoxic and mutagenic properties in mouse lymphoma cells.
  • Acts as a nitric oxide radical
  • Functions as a selective scavenger of reactive oxygen species (ROS) in mitochondria
  • Serves as an organocatalyst, disproportionating superoxide
  • Oxidizes primary alcohols to aldehydes in a catalytic cycle
  • Exhibits mutagenic and antioxidant effects
  • Induces DNA strand breaks
  • Exerts cytotoxic and mutagenic properties in mouse lymphoma cells
Catalog No. 50-004-46968