Learn More
Medchemexpress LLC Tempol (4-hydroxy-2,2,6,6-tetramethylpiperidin-1-oxyl) | 2226-96-2 | MFCD00006478 | 99.7% | 172.24 g/mol | C9H18NO2 | 10 G
SDP
Supplier:  Medchemexpress LLC HY10056110G
Encompass_Preferred
Tempol (4-hydroxy-2,2,6,6-tetramethylpiperidin-1-oxyl) is a stable nitroxide radical used in research as a superoxide dismutase (SOD)-mimetic to scavenge reactive oxygen species (ROS). It is provided as a high-purity reagent for laboratory and analytical applications, and is available in solid and solution formats to support different experimental workflows.
  • Stable nitroxide radical suitable for antioxidant studies.
  • Mimics superoxide dismutase activity to neutralize ROS.
  • High purity for reproducible experimental results.
  • Available as solid and solution formulations for flexibility.
  • Molecular weight 172.24 g/mol and formula C9H18NO2.
  • Intended for research and analytical use only, not for human use.
Catalog No. 50-002-12690