Learn More
Medchemexpress LLC Tempol-d17,15N | 90429-66-6 | 98.4% | 190.34 g/mol | C9HD1715NO2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY100561S5MG
Encompass_Preferred
Tempol-d17,15N is a deuterium- and nitrogen-15-labeled analogue of 4-hydroxy-TEMPO, a superoxide dismutase (SOD)-mimetic that neutralizes reactive oxygen species. The isotopic labels provide a predictable mass shift for tracer studies and analytical quantification, enabling mechanistic and mass spectrometry experiments at microgram scales.
  • Isotopically labeled with deuterium (d17) and 15N for mass-shifted detection.
  • Suitable for tracer and analytical mass spectrometry applications.
  • High listed purity for reliable experimental results.
  • Small, microgram-scale packaging ideal for analytical workflows.
  • Applicable to studies of reactive oxygen species and SOD-mimetic activity.
Catalog No. 50-002-13078