Learn More
USA Scientific Inc TempPlate semskrt PP 0.1 mL
Supplier:  USA Scientific Inc 14028990
Encompass
TempPlate semskrted polypropylene 0.1 mL 96-well PCR plate, low profile, straight sides, white, 10/box
Catalog No. 50-274-5545