Learn More
Medchemexpress LLC HY-50910 25mg , Temsirolimus CAS:162635-04-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY5091025MG
Encompass_Preferred
HY-50910 25mg Temsirolimus CAS: 162635-04-3 Temsirolimus is an inhibitor of mTOR with an IC50 of 1.76 μM. Temsirolimus activates autophagy and prevents deterioration of cardiac function in animal model[8]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9567