Learn More
Selleck Chemical LLC Temsirolimus (CCI-779) 200mg 162635-04-3 NSC 683864
SDP
Supplier:  Selleck Chemical LLC S1044200MG
Encompass_Preferred
Temsirolimus (CCI-779, NSC 683864) is a specific mTOR inhibitor with IC50 of 1.76 ?M in a cell-free assay. Temsirolimus induces autophagy and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-811-9