Learn More
Sigma Aldrich Fine Chemicals Biosciences Ten Eleven Translocase TET
Supplier:  Sigma Aldrich Fine Chemicals Biosciences S979725UG
Encompass
This product consists of the catalytic domain (CD) of mTET1 (amino acids 1366-2039) and includes an N-terminal polyhistidine tag. It has a calculated molecular mass of 75.8 kDa. This protein is produced in mammalian cells (HEK 293) without the use of serum. The mammalian cells expression system allows proper post-translational modifications and folding and often supports higher specific activity of the protein.
Catalog No. 50-290-4405