Learn More
Selleck Chemical LLC Tenalisib (RP6530) 2mg 1639417-53-0
SDP
Supplier:  Selleck Chemical LLC S86722MG
Encompass_Preferred
Tenalisib (RP6530) is a potent and selective dual PI3K?/? inhibitor with IC50 values of 24.5 nM and 33.2 nM for PI3K? and PI3K?, respectively. Its selectivity over ? and ? isoforms are more than 300-fold and 100-fold, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8336