Learn More
Medchemexpress LLC Teneligliptin | 760937-92-6 | 99.8% | 426.58 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY14806
Encompass_Preferred
Teneligliptin hydrobromide hydrate is an orally active and selective dipeptidyl peptidase 4 (DPP-4) inhibitor with IC50s of 0.37 and 0.29 nM for human and rat DPP-4, respectively. It improves blood glucose levels and is used in research related to type 2 diabetes mellitus. It is for research use only and not sold to patients.
  • Orally active and selective dipeptidyl peptidase 4 (DPP-4) inhibitor
  • Improves blood glucose levels
  • Used in research related to type 2 diabetes mellitus
  • For research use only
Catalog No. 50-003-63457