Learn More
Medchemexpress LLC HY-14806A 10mg Medchemexpress, Teneligliptin (hydrobromide) CAS:906093-29-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY14806A10MG
Encompass_Preferred
Medchemexpress, HY-14806A 10mg Teneligliptin (hydrobromide) CAS:906093-29-6 Teneligliptin (MP-513) hydrobromide is a potent chemotype prolylthiazolidine-based DPP-4 inhibitor, which competitively inhibits human plasma, rat plasma, and human recombinant DPP-4 in vitro, with IC50s of approximately 1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1643