Learn More
Enterprise Technology Solutions Tennsco Corp. HighStorageCabinet,Jumbo,5Shelf,400lbCap,48"x18"x78",MedGrey
SDP
Supplier:  Enterprise Technology Solutions TNNJ1878SUMGY
Encompass_Preferred
Jumbo heavy-gauge steel cabinet offers extra capacity for big storage problems. Specially engineered shelves utilize box-formed construction to accommodate heavy loads even with the extra-wide shelf span. Four of the five shelves adjust in 2" increments without tools. Built-in shelf support tabs secure shelves tightly to the cabinet without clips or added parts, adding rigidity and preventing small items from falling over edge or rear of shelf. Dual swing-out doors ride on three heavy-duty leaf-hinges to assure doors stay aligned. Each door incorporates an extra-wide reinforcement that is welded for added strength and stability. Two brushed chrome handles, with one that locks both doors, make door opening and closing easy. With three-point locking mechanism, doors lock at three points for maximum security. Nylon guides help make operation quiet. The raised base keeps materials out of harm's way from cleaning liquids. Two adjustable levelers compensate for uneven floors.
Catalog No. 50-007-91134