Learn More
Selleck Chemical LLC Tenofovir S1401-1g
SDP
Supplier:  Selleck Chemical LLC S14011G
Encompass_Preferred
Tenofovir (GS-1278) blocks reverse transcriptase and hepatitis B virus infections
Catalog No. 50-311-0293