Learn More
Medchemexpress LLC HY-15232 10mg , Tenofovir alafenamide CAS:379270-37-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1523210MG
Encompass_Preferred
HY-15232 10mg Tenofovir alafenamide CAS: 379270-37-8 Tenofovir alafenamide (GS-7340) is an investigational oral prodrug of Tenofovir. Tenofovir is a HIV-1 nucleotide reverse transcriptase inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0151