Learn More
Medchemexpress LLC Tenofovir alafenamide fumarate | 379270-38-9 | C25H33N6O9P | 100 MG
SDP
Supplier:  Medchemexpress LLC HY15232A100MG
Encompass_Preferred
Tenofovir alafenamide fumarate (GS-7340 fumarate) is an investigational oral prodrug of Tenofovir, an HIV-1 nucleotide reverse transcriptase inhibitor. It demonstrates antiviral activities against HIV-1 and HIV-2 isolates, with EC50s ranging from 0.1 to 12 nM for primary HIV-1 isolates. This amidate prodrug of Tenofovir offers good oral bioavailability and increased plasma stability compared to Tenofovir disoproxil fumarate (TDF).
  • Investigational oral prodrug of Tenofovir
  • HIV-1 nucleotide reverse transcriptase inhibitor
  • Antiviral activities against HIV-1 and HIV-2 isolates
  • Good oral bioavailability
  • Increased plasma stability
Catalog No. 50-003-73885