Learn More
Medchemexpress LLC Tenofovir alafenamide fumarate (GS-7340 fumarate) | 379270-38-9 | C25H33N6O9P | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15232A10MM/1ML
Encompass_Preferred
Tenofovir alafenamide fumarate is an investigational oral prodrug of Tenofovir, functioning as an HIV-1 nucleotide reverse transcriptase inhibitor. It is intended solely for research purposes and not for patient use.
  • Exhibits antiviral activities ranging from 5 to 7 nM
  • Effective against a panel of HIV-1 and HIV-2 isolates
  • Mean EC50 of 3.5 nM against primary HIV-1 isolates in PBMCs
  • Mean EC50 of 1.8 nM for HIV-2 isolates
  • Amidate prodrug designed for improved oral bioavailability
  • Offers increased plasma stability compared to Tenofovir disoproxil fumarate (TDF)
  • High purity of 99.87%
  • Appears as a white to off-white solid
  • Soluble in DMSO (≥ 36 mg/mL) and H2O (≥ 25 mg/mL)
  • Store solid form at 4°C, sealed and away from moisture
  • Store solvent form at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-003-73184