Learn More
Medchemexpress LLC Tenofovir alafenamide fumarate (GS-7340 fumarate) | 379270-38-9 | C25H33N6O9P | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15232A50MG
Encompass_Preferred
Tenofovir alafenamide fumarate is an investigational oral prodrug of Tenofovir, an HIV-1 nucleotide reverse transcriptase inhibitor. It offers good oral bioavailability and increased plasma stability compared to Tenofovir disoproxil fumarate (TDF). This product is for research use only and not sold to patients.
  • Active against HIV-1 and HIV-2 isolates
  • Similar antiviral activities across various cell types
  • Effective against HIV-1 group M subtypes A to G, and group N and O isolates
Catalog No. 50-003-73104