Learn More
Medchemexpress LLC Tenofovir alafenamide hemifumarate | 1392275-56-7 | C21H29N6O5P.1/2 C4H4O4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15232B10MM/1ML
Encompass_Preferred
Tenofovir alafenamide hemifumarate (GS-7340 hemifumarate) is an investigational oral proagent of Tenofovir. Tenofovir is an HIV-1 nucleotide reverse transcriptase inhibitor.
  • Investigational oral proagent
  • HIV-1 nucleotide reverse transcriptase inhibitor
  • Amidate prodrug with good oral bioavailability
  • Increased plasma stability
Catalog No. 50-003-73548