Learn More
Medchemexpress LLC Tenofovir alafenamide hemifumarate | 1392275-56-7 | C21H29N6O5P.1/2 C4H4O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15232B50MG
Encompass_Preferred
Tenofovir alafenamide hemifumarate is an investigational oral prodrug of Tenofovir, an HIV-1 nucleotide reverse transcriptase inhibitor. It offers good oral bioavailability and increased plasma stability compared to Tenofovir disoproxil fumarate (TDF).
  • Antiviral activities range from 5 to 7 nM across all cell types
  • CC50 varies from 4.7 to 42 μM for MT-4 and MT-2 cells, respectively
  • Effective against a panel of HIV-1 and HIV-2 isolates
  • Mean EC50 for 29 primary HIV-1 isolates in PBMCs is 3.5 nM
  • Mean EC50 for HIV-2 isolates is 1.8 nM
Catalog No. 50-003-73389