Learn More
Medchemexpress LLC Tenofovir amibufenamide | 1571076-26-0 | 99.7% | 490.49 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY13745310MM/1ML
Encompass_Preferred
Tenofovir amibufenamide is a Tenofovir prodrug and an orally active antiviral agent for research use. It inhibits HBV and can be used for the study of chronic hepatitis B (CHB).
  • Inhibits HBV DNA replication with an EC50 of 7.3 nM in HepG2.2.15 cells.
  • Corrects metabolic disorders in host liver cells caused by HBV infection.
  • Regulates hepatocyte metabolism and exhibits antiviral effects in HBV transgenic mouse models.
  • Decreases HBV DNA levels in serum.
Catalog No. 50-003-51785