Learn More
Medchemexpress LLC Tenofovir amibufenamide | 1571076-26-0 | 99.7% | 490.49 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHY137453
Encompass_Preferred
Tenofovir amibufenamide is a Tenofovir prodrug and an orally active antiviral agent. It inhibits HBV and is used for chronic hepatitis B (CHB) studies.
  • Inhibits HBV DNA replication in HepG2.2.15 cells (EC50 of 7.3 nM).
  • Corrects metabolic disorders in host liver cells caused by HBV infection.
  • Regulates hepatocyte metabolism in HBV transgenic mice.
  • Decreases HBV DNA levels in serum in HBV transgenic mice.
Catalog No. 50-003-64190