Learn More
TARGETMOL CHEMICALS INC TENOFOVIR AMIBUFENAMIDE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T390435MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Tenofovir amibufenamide (HS-10234) (HS-10234) is a Tenofovir prodrug an antiviral compound with oral activity. Tenofovir amibufenamide inhibits hepatitis B virus (HBV) and can be used in chronic hepatitis B (CHB) studies. purity: 99%
Catalog No. 50-258-0071